Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Heptane/Toluene, 90/10 v/v, Reagents

Catalog No. 01335148
Change view
Click to view available options
:
Quantity: 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-335-148 Quantity: 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-335-148 Supplier Reagents Holdings Llc Supplier No. CH0040004C
Only null left
Add to Cart
Add to Cart

n-Heptane/Toluene, 90/10 v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 108-88-3 , 142-82-5
Molecular Formula C7H8
Molecular Weight (g/mol) 92.14
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
Synonym Dipropylmethane/Toluol
IUPAC Name toluene
SMILES CC1=CC=CC=C1

Specifications

Color Colorless
Quantity 4 L
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 0.6994
Grade Reagent
Chemical Name or Material n-Heptane/Toluene, 90/10 v/v
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.