Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexyl alcohol, 98%, pure

Catalog No. AC120791000
Click to view available options
Quantity:
100 mL
1 L
2.5 L
10 L
25 L
Packaging:
Glass bottle
Metal Drum
Plastic drum
This item is not returnable. View return policy

Catalog No. AC120791000

Supplier: Thermo Scientific Chemicals 120791000

Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 111-27-3 | C6H14O | 102.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-27-3
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.18
MDL Number MFCD00002982
InChI Key ZSIAUFGUXNUGDI-UHFFFAOYSA-N
Synonym 1-hexanol, hexyl alcohol, hexanol, n-hexanol, n-hexyl alcohol, 1-hexyl alcohol, 1-hydroxyhexane, amylcarbinol, caproyl alcohol, pentylcarbinol
PubChem CID 8103
ChEBI CHEBI:87393
IUPAC Name hexan-1-ol
SMILES CCCCCCO

Specifications

Acidity 0.02mg KOH/g max.
Melting Point -52.0°C
Density 0.8136g/mL
Boiling Point 156.0°C to 157.0°C
Flash Point 59°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4170 to 1.4190
Linear Formula CH3(CH2)5OH
Quantity 100 mL
Beilstein 01, 407
Specific Gravity 0.8136
Merck Index 15, 4734
Solubility Information Solubility in water: 6g/L (25°C). Other solubilities: miscible with alcohol and ether
Viscosity 5.3 mPa.s (20°C)
Formula Weight 102.18
Percent Purity 98%
Grade Pure
Physical Form Liquid
Chemical Name or Material Hexyl alcohol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously wit
EINECSNumber 203-852-3

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.