Learn More
n-Hexane, 97%, Acros Organics™
Specifications
Linear Formula | CH3(CH2)4CH3 |
Solubility Information | Solubility in water: insoluble. Other solubilities: soluble in alcohol, acetone, ether and chloroform |
Formula Weight | 86.18 |
Percent Purity | 97% |
Grade | Extra Dry over Molecular Sieve |
Density | 0.6590g/mL |
Refractive Index | 1.3748 to 1.3810 |
Solubility | Solubility in water: insoluble. Other solubilities: soluble in alcohol, acetone, ether and chloroform |
Specific Gravity | 0.659 |
Water | 0.005% max. (Coulometric) |
Show More |
Safety and Handling
GHS H Statement May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. Causes skin irritation. May cause damage to organs through prolonged or repeated exposure. Toxic to aquatic life with long lasting effects. Suspected of damaging fertility. Highly flammable liquid and vapor.
GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Use personal protective equipment as required. Do not breathe dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. IF ON SKIN: Wash with plenty of soap and water.
Danger