Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Hexane, anhydrous

Catalog No. aa43263ak
Change view
Click to view available options
Quantity:
250 mL
1 L
4 L
Catalog No. Quantity
AA43263AK 250 mL
AA43263K2 1 L
AA43263K7 4 L
3 options

Catalog No. AA43263AK

Supplier: Thermo Scientific Chemicals 043263AK

Only null left
Add to Cart

Solvent in Sol-Gel Precursor Applications | CAS: 110-54-3 | C6H14 | 86.18 g/mol

n-Hexane is used as a solvent to extract edible oils from seed and vegetable crops. Commercial grades of hexane are used as solvents for glues, varnishes, and inks, cleaning agent (degreaser) in the printing industry, liquid in low temperature thermometers. Other uses are as fuels and fuel additives, intermediates and laboratory chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC

Specifications

Melting Point °C to 95°C
Color Colorless
Density 0.659 g/cm3 at 20°C
Boiling Point 69°C
Quantity 250 mL
Flash Point -23°C (-9°F)
Ignition Point 240°C (464°F)
Odor Weak, characteristic
Physical Form Liquid
Refractive Index 1.375
Vapor Pressure 120mm Hg at 20°C
UN Number 1208
Beilstein 1730733
Solubility Information Miscible with alcohol,chloroform,acetone and ether.
Formula Weight 86.18
Chemical Name or Material n-Hexane
Show More Show Less
Hazard Category H225-H304-H315-H319-H335-H336-H361f-H372
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280i-P281-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation DOT Class: 3, Packaging Group: II
EINECSNumber 203-777-6
RTECSNumber MN9275000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.