Learn More
n-Hexane, anhydrous, over molecular sieves, packaged under argon in resealable ChemSeal™ bottles
Description
n-Hexane is used as a solvent, paint thinner, and chemical reaction medium. It is used to extract edible oils from seeds and vegetables, as a special-use solvent, and as a cleaning agent. Hexanes are commonly used in chromatography as a non-polar solvent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under Argon

Specifications
Specifications
Melting Point | -95°C |
Color | Colorless |
Density | 0.659 g/mL |
Boiling Point | 69°C |
Quantity | 250 mL |
Flash Point | -23°C (-9°F) |
Packaging | ChemSeal™ Bottle |
Physical Form | Liquid |
Refractive Index | 1.375 |
Linear Formula | CH3(CH2)4CH3 |
Show More |
Safety and Handling
Hazard Category | H225-H304-H315-H319-H335-H336-H361f-H372 |
Hazard Statement | GHS H Statement H225-H304-H361-H373-H315-H336 Highly flammable liquid and vapor. May be fatal if swallowed and enters airways. Suspected of damaging fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Causes skin irritation. May cause drowsiness or dizziness. |
Precautionary Statement | P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280i-P281-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P331-P332+P313-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: HEXANES |
EINECSNumber | 203-777-6 |
RTECSNumber | MN9275000 |
TSCA | Yes |
storageNote1 |
|
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.