Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Hexane For gas chromatography ECD and FID SupraSolv™, MilliporeSigma™

Catalog No. m1043711000
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle

For gas chromatography ECD and FID SupraSolv™

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC

Specifications

Melting Point -94.3°C
Color Colorless
Density 0.66g/cm3 at 20°C
Boiling Point 69°C
Quantity 1 L
Flash Point −22°C (−7.6°F)
Ignition Point 240° (464°F)
Odor Benzine-like
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.37
Vapor Pressure 160 hPa (20°C)
UN Number 1208
Solubility Information Soluble in water: 0.0095g/L at 20°C
Formula Weight 86.18
Chemical Name or Material Hexane
Show More Show Less
Hazard Category H225: Highly flammable liquid and vapour.
Hazard Statement H304: May be fatal if swallowed and enters airways.
Precautionary Statement H315: Causes skin irritation.
Recommended Storage 5°C to 30°C (41°F to 86°F); Keep container tightly closed in a dry and well-ventilated place. Keep away from heat and sources of ignition.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.