Learn More
n-Hexane, HPLC Grade, 95% min
Description
as a solvent to extract edible oils from seed and vegetable crops. Commercial grades of hexane are used as solvents for glues, varnishes, and inks, cleaning agent (degreaser) in the printing industry, liquid in low temperature thermometers. other uses are as Fuels and fuel additives, Intermediates and Laboratory chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
            Specifications
Specifications
| Melting Point | °C to 95°C | 
| Density | 0.659 | 
| Boiling Point | 69°C | 
| Quantity | 1 L | 
| Flash Point | -23°C (-9°F) | 
| Physical Form | Liquid, Distilled in Glass | 
| Refractive Index | 1.375 | 
| Assay Percent Range | 95% min | 
| Beilstein | 1730733 | 
| Solubility Information | Very soluble in ethanol,ethyl ether and chloroform. | 
| Show More | 
Safety and Handling
| missing translation for 'healthHazard1' | H225-H304-H315-H319-H335-H336-H361f-H372 | 
| missing translation for 'healthHazard3' | P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280i-P281-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P331-P332+P313-P363-P370+P378q-P501c | 
| DOTInformation | Hazard Class: 3; Packaging Group: II | 
| EINECSNumber | 203-777-6 | 
| RTECSNumber | MN9275000 | 
| TSCA | Yes | 
| Recommended Storage | Ambient temperatures | 
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.