Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(Hydroxymethyl)acetamide, 98+%, Thermo Scientific™

Catalog No. p-7049939
Click to view available options
Quantity:
5 g
100 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 625-51-4
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.09
MDL Number MFCD00014417
InChI Key HWJHZLJIIWOTGZ-UHFFFAOYSA-N
Synonym n-hydroxymethyl acetamide, acetamidomethanol, formicin, acetamide, n-hydroxymethyl, unii-rcg03a51mo, n-hydroxymethyl-acetamide, rcg03a51mo, acetamido methanol, n-acetylmethanolamine, acetylamino methanol
PubChem CID 69365
IUPAC Name N-(hydroxymethyl)acetamide
SMILES CC(=O)NCO

Specifications

Melting Point 55°C to 56°C
Quantity 5 g
Formula Weight 89.1
Percent Purity ≥98%
Physical Form Crystalline
Chemical Name or Material N-(Hydroxymethyl)acetamide
Hazard Category Danger
Hazard Statement GHS H Statement
H311-H332-H319
Toxic in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.