Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium perchlorate monohydrate, 85+%, ACS reagent
Click to view available options
Quantity:
100 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent

Specifications
Specifications
Melting Point | 130.0°C |
Color | White to Yellow |
pH | 6.0 to 8.0 (5% aq. soln. at 25°C) |
Physical Form | Crystalline Powder |
Quantity | 100 g |
Assay Percent Range | 85.0 to 90% (as NaClO4) (Argentometry) |
Molecular Formula | ClNaO4·H2O |
Linear Formula | ClNaO4·H2O |
Merck Index | 15,8784 |
Solubility Information | Solubility in water: 2090g/L (15°C). Other solubilities: soluble in alcohol |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement May cause fire or explosion; strong oxidizer. Harmful if swallowed. |
Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Take any precaution to avoid mixing with combustibles. |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction