Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Hydroxyphthalimide, 98%

Catalog No. p-147599
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Plastic drum

CAS: 524-38-9 | C8H5NO3 | 163.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 524-38-9
Molecular Formula C8H5NO3
Molecular Weight (g/mol) 163.13
MDL Number MFCD00005891
InChI Key CFMZSMGAMPBRBE-UHFFFAOYSA-N
Synonym n-hydroxyphthalimide, 2-hydroxyisoindoline-1,3-dione, 2-hydroxy-1h-isoindole-1,3 2h-dione, nhpi, 2-hydroxyphthalimide, 1h-isoindole-1,3 2h-dione, 2-hydroxy, phthalimide, n-hydroxy, phthaloxime, unii-bxi99m81x0, 2-hydroxyisoindolin-1,3-dione
PubChem CID 10665
IUPAC Name 2-hydroxyisoindole-1,3-dione
SMILES ON1C(=O)C2=CC=CC=C2C1=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 233.0°C to 235.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Quantity 100 g
Beilstein ,5
Fieser 01,485; 07,177
Merck Index 15,4876
Solubility Information Solubility in water: slightly soluble
Formula Weight 163.13
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N-Hydroxyphthalimide, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
EINECSNumber 208-358-1
RTECSNumber TI5200000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.