Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Iodosuccinimide, 97%

Catalog No. p-3346177
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 516-12-1 | C4H4INO2 | 224.99 g/mol

N-Iodosuccinimide is used in the preparation of vinyl sulfones from olefins and benzenesulfinic acid. It acts as a source for I+ and involved in Hunsdiecker reactions for the conversion of cinnamic acids, and propiolic acids to the corresponding alfa-halostyrenes and 1-halo-1-alkynes respectively. It is also used to hydrolyze thioglycosides to 1-hydroxyglycosides with triluoroacetic acid. It is involved in the preparation of iodobenzene from 1,6-diynes. Further, it acts as an iodinating agent in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Iodosuccinimide is used in the preparation of vinyl sulfones from olefins and benzenesulfinic acid. It acts as a source for I+ and involved in Hunsdiecker reactions for the conversion of cinnamic acids, and propiolic acids to the corresponding alfa-halostyrenes and 1-halo-1-alkynes respectively. It is also used to hydrolyze thioglycosides to 1-hydroxyglycosides with triluoroacetic acid. It is involved in the preparation of iodobenzene from 1,6-diynes. Further, it acts as an iodinating agent in chemical synthesis.

Solubility
Soluble in dioxane, tetrahydrfuran and acetonitrile. Insoluble in ether and carbon tetrachloride.

Notes
Moisture and light sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 516-12-1
Molecular Formula C4H4INO2
Molecular Weight (g/mol) 224.99
MDL Number MFCD00005512
InChI Key LQZMLBORDGWNPD-UHFFFAOYSA-N
Synonym n-iodosuccinimide, succiniodimide, n-iodo succinimide, 2,5-pyrrolidinedione, 1-iodo, unii-3cos3x3n4p, nis, succinimide, n-iodo, 3cos3x3n4p, 1-iodo-pyrrolidine-2,5-dione, 1-iodoazolidine-2,5-dione
PubChem CID 120273
ChEBI CHEBI:53204
IUPAC Name 1-iodopyrrolidine-2,5-dione
SMILES IN1C(=O)CCC1=O

Specifications

Melting Point ∽196°C (decomposition)
Density 2.245
Assay Percent Range 97%
Quantity 5 g
Beilstein 113917
Sensitivity Light and moisture sensitive
Solubility Information Soluble in dioxane,tetrahydrfuran and acetonitrile. Insoluble in ether and carbon tetrachloride.
Formula Weight 224.99
Chemical Name or Material N-Iodosuccinimide
Hazard Category H302-H315-H319-H335-H500
Precautionary Statement P261-P264b-P270-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 208-221-6
RTECSNumber WN2817000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.