Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Iodosuccinimide 98.0+%, TCI America™
SDP

Catalog No. I0074100G
Click to view available options
Quantity:
5 g
25 g
100 g

Chemical Identifiers

CAS 516-12-1
Molecular Formula C4H4INO2
Molecular Weight (g/mol) 224.99
MDL Number MFCD00005512
InChI Key LQZMLBORDGWNPD-UHFFFAOYSA-N
Synonym n-iodosuccinimide, succiniodimide, n-iodo succinimide, 2,5-pyrrolidinedione, 1-iodo, unii-3cos3x3n4p, nis, succinimide, n-iodo, 3cos3x3n4p, 1-iodo-pyrrolidine-2,5-dione, 1-iodoazolidine-2,5-dione
PubChem CID 120273
ChEBI CHEBI:53204
IUPAC Name 1-iodopyrrolidine-2,5-dione
SMILES IN1C(=O)CCC1=O

Specifications

Melting Point 201°C
Quantity 100 g
Formula Weight 224.99
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Iodosuccinimide
RTECSNumber WN2817000
TSCA No
Recommended Storage Freezer
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.