Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Isopropylacrylamide, 97%

Click to view available options
Quantity:
25g
100g
2210-25-5
C6H11NO
113.16
MFCD00041913
QNILTEGFHQSKFF-UHFFFAOYSA-N
n-isopropylacrylamide,nipam,n-iso-propylacrylamide,2-propenamide, n-1-methylethyl,isopropyl acrylamide,acrylamide, n-isopropyl,n-isopropyl acrylamide,n-1-methylethyl-2-propenamide,isopropylamid kyseliny akrylove,unii-b7gff17l9u
16637
N-propan-2-ylprop-2-enamide
CC(C)NC(=O)C=C
2210-25-5
C6H11NO
113.16
MFCD00041913
QNILTEGFHQSKFF-UHFFFAOYSA-N
n-isopropylacrylamide,nipam,n-iso-propylacrylamide,2-propenamide, n-1-methylethyl,isopropyl acrylamide,acrylamide, n-isopropyl,n-isopropyl acrylamide,n-1-methylethyl-2-propenamide,isopropylamid kyseliny akrylove,unii-b7gff17l9u
16637
N-propan-2-ylprop-2-enamide
CC(C)NC(=O)C=C
Applications
N-Isopropylacrylamide is used to produce poly(N-isopropylacrylamide) (pNIPA, pNIPAAm, pNIPAA or pNIPAm) thermo sensitive polymer- or copolymer-based hydrogels. Polymers that contain NIPAM shrink dramatically above 33°C. Monomer used in the preparation of thermally sensitive, water-swellable hydrogels.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Quantity 25g
Boiling Point 89°C to 92°C (2mmHg)
Melting Point 60°C to 63°C
Beilstein 1744926
Formula Weight 113.16
Physical Form Solid
Percent Purity 97%
Chemical Name or Material N-Isopropylacrylamide

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : AS3675000

TSCA : TSCA

storageNote1 : 2° to 8°C