Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Isopropylmethylamine, 98%

Click to view available options
Quantity:
5g
25g
4747-21-1
C4H11N
73.139
MFCD00042859
XHFGWHUWQXTGAT-UHFFFAOYSA-N
n-isopropylmethylamine,n-methylisopropylamine,methylisopropylamine,2-propanamine, n-methyl,isopropylmethylamine,2-methylaminopropane,methyl propan-2-yl amine,ethylamine, n,1-dimethyl,unii-j3ol90426g,n-methylisopropyylamine
78485
N-methylpropan-2-amine
CC(C)NC
4747-21-1
C4H11N
73.139
MFCD00042859
XHFGWHUWQXTGAT-UHFFFAOYSA-N
n-isopropylmethylamine,n-methylisopropylamine,methylisopropylamine,2-propanamine, n-methyl,isopropylmethylamine,2-methylaminopropane,methyl propan-2-yl amine,ethylamine, n,1-dimethyl,unii-j3ol90426g,n-methylisopropyylamine
78485
N-methylpropan-2-amine
CC(C)NC
Applications
N-Isopropylmethylamine is used in the generation of in situ micelles having distinct sizes, by sequential self-assembly of a series of stimuli-responsive amphiphilic block copolymers, prepared by reversible addition-fragmentation chain transfer (RAFT) polymerization. It is suitable for use in the synthesis of novel 5-ethoxy-N,N-dialkyl-[α,α,β,β-H4]-tryptamines and their [α,α,β,β,β-D4]-counterparts following the Speeter and Anthony procedure.

Solubility
Soluble in chloroform, DMSO, methanol.

Notes
Air Sensitive. Store away from air. Store under dry inert gas. Do not store together with acids. Incompatible with acids , air and oxidizing agents.
Boiling Point 50°C to 53°C
Quantity 5g
Sensitivity Air Sensitive
Refractive Index 1.3480
Color Colorless
Density 0.702g/mL
Linear Formula (CH3)2CHNHCH3
UN Number UN2734
Beilstein 1730877
Formula Weight 73.14
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material N-Isopropylmethylamine
Show More Show Less

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packaging Group: I; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, FLAMMABLE, N.O.S.