Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
N-(-Ketocaproyl)-DL-homoserine lactone, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
25 mg
50 mg
Description
Featured Industries: Pharmaceutical (small molecule)
Specifications
Specifications
| CAS | 76924-95-3 |
| Physical Form | Neat |
| Linear Formula | C10H15 NO4 |
| MDL Number | MFCD03788849 |
| Synonym | N-(3-Oxohexanoyl)-DL-homoserine lactone |
| Type | Analytical Standard |
| Quantity | 50 mg |
| Formula Weight | 213.23 |
| Grade | Analytical Standard |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction