Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Lauroyl-D-erythro-sphingosine, ≥99%, synthetical, Thermo Scientific™

Catalog No. AC346250500
Click to view available options
Quantity:
50 mg

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74713-60-3
Molecular Formula C30H59NO3
Molecular Weight (g/mol) 481.806
MDL Number MFCD02259133
InChI Key HXFPPRPLRSPNIB-VARSQMIESA-N
Synonym laurylsphingosine, dodecyl sphingosine, c12 ceramide, n-lauroylsphingosine, n-dodecanoyl-ceramide, n-lauroyl-d-erythro-sphingosine, n-dodecanoyl-sphing-4-enine, n-dodecanoylsphingosine, n-lauroyl ceramide, n-dodecanoyl ceramide
PubChem CID 5283562
ChEBI CHEBI:72956
IUPAC Name N-[(E,2S,3R)-1,3-dihydroxyoctadec-4-en-2-yl]dodecanamide
SMILES CCCCCCCCCCCCCC=CC(C(CO)NC(=O)CCCCCCCCCCC)O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Quantity 50 mg
Formula Weight 481.8
Percent Purity ≥99.0%
Physical Form Powder
Chemical Name or Material N-Lauroyl-D-erythro-sphingosine, 99+%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.