Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Me-6,7-diOH-TIQ Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB167SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB167SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB167SP Supplier R&D Systems Supplier No. MAB167SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

N-Me-6,7-diOH-TIQ Monoclonal specifically detects N-Me-6,7-diOH-TIQ in Multi-Species samples. It is validated for Immunohistochemistry.

Specifications

Antigen N-Me-6,7-diOH-TIQ
Applications Immunohistochemistry
Classification Monoclonal
Clone 106814
Conjugate Unconjugated
Dilution Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Host Species Mouse
Immunogen Carrier protein-coupled N-Me-6,7-diOH-TIQ
Purification Method Protein A or G purified from ascites
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes N-Me-6,7-diOH-TIQ in immunoassays. The specificity was determined by a competition type assay where the N-Me-6,7-diOH-TIQ labeled with biotin at position 5 or 8 competed with another compound for binding to the antibody binding site. The detection limit of biotinylated N-Me-6,7-diOH-TIQ in assays where the molecule was captured by the antibody coated on microplates and developed by HRP-streptavidin was 5 nM. The dissociation constants of the interaction between the antibody and N-Me-6,7-diOH-TIQ and its biotinylated analog were 0.2 and 0.5 μM, respectively. This antibody specifically recognizes N-Me-6,7-diOH-TIQ and shows 60% cross-reactivity with 6,7-diOH-TIQ, 15% cross-reactivity with 1,N-di-Me-6,7-diOH-TIQ, 3% cross-reactivity with 1-Me-6,7-diOH-TIQ, and less than 1% or no cross-reactivity with tetrahydroisoquinoline (TIQ), 5-OH-isoquiniline, 5-OH-N-Me-isoquinolinium, tetrahydropapaveroline, 9H-pyrido[3,4-b]indole, and 1,2,3,4-tetrahydro-9H-pyrido[3,4-b]indole. The percent cross-reactivity is expressed by the relative value, which produced 50% inhibition of interaction between the biotin-labeled N-methyl-6,7-dihydroxytetrahydroisoquinoline and the other compound.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Multi-species
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.