Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methyl-4-nitroaniline, 97%, Thermo Scientific Chemicals

Catalog No. AAA1554814
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1554814 25 g
AAA1554806 5 g
AAA1554822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1554814 Supplier Thermo Scientific Chemicals Supplier No. A1554814
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-15-2
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.15
MDL Number MFCD00007305
InChI Key XIFJZJPMHNUGRA-UHFFFAOYSA-N
Synonym n-methyl-p-nitroaniline, benzenamine, n-methyl-4-nitro, 4-nitro-n-methylaniline, n-methyl-p-nitraniline, p-nitro-n-methylaniline, p-methylamino nitrobenzene, n-monomethyl-p-nitroaniline, aniline, n-methyl-p-nitro, 4-methylaminonitrobenzene, methyl 4-nitrophenyl amine
PubChem CID 7483
IUPAC Name N-methyl-4-nitroaniline
SMILES CNC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 147°C to 154°C
Odor Characteristic
Quantity 25 g
UN Number UN2811
Beilstein 637920
Formula Weight 152.15
Percent Purity 97%
Chemical Name or Material N-Methyl-4-nitroaniline
Hazard Category H301+H311+H331-H315-H319-H335-H500
Hazard Statement GHS H Statement
H301-H311-H331-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 202-823-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.