Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-methyl-bis(trifluoroacetamide), 98%, ACROS Organics™

Click to view available options
Quantity:
5mL
25mL
Packaging:
Glass bottle
685-27-8
C5H3F6NO2
223.07
AWGBWLXGUPTXHF-UHFFFAOYSA-N
mbtfa, n-methyl-bis trifluoroacetamide, n-methylbis trifluoroacetamide, 2,2,2-trifluoro-n-methyl-n-trifluoroacetyl acetamide, n,n-bis trifluoroacetyl methylamine, 2,2,2-trifluoro-n-methyl-n-2,2,2-trifluoroacetyl acetamide, acetamide, 2,2,2-trifluoro-n-methyl-n-trifluoroacetyl, n-methylbistrifluoroacetamide, n-methyl-bis-trifluoroacetamide
69635
2,2,2-trifluoro-N-methyl-N-(2,2,2-trifluoroacetyl)acetamide
CN(C(=O)C(F)(F)F)C(=O)C(F)(F)F
685-27-8
C5H3F6NO2
223.07
AWGBWLXGUPTXHF-UHFFFAOYSA-N
mbtfa, n-methyl-bis trifluoroacetamide, n-methylbis trifluoroacetamide, 2,2,2-trifluoro-n-methyl-n-trifluoroacetyl acetamide, n,n-bis trifluoroacetyl methylamine, 2,2,2-trifluoro-n-methyl-n-2,2,2-trifluoroacetyl acetamide, acetamide, 2,2,2-trifluoro-n-methyl-n-trifluoroacetyl, n-methylbistrifluoroacetamide, n-methyl-bis-trifluoroacetamide
69635
2,2,2-trifluoro-N-methyl-N-(2,2,2-trifluoroacetyl)acetamide
CN(C(=O)C(F)(F)F)C(=O)C(F)(F)F
Quantity 5mL
Boiling Point 120°C to 122°C
Specific Gravity 1.547
Flash Point 45°C
Density 1.5470g/mL
Packaging Glass bottle
Assay Percent Range 97.5% min. (GC)
Linear Formula (CF3CO)2NCH3
Formula Weight 223.07
Percent Purity 98%
Chemical Name or Material N-Methyl-bis(trifluoroacetamide)
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapor.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 211-680-5