Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methyl-D-phenylalanine, 97%, Thermo Scientific™

Catalog No. AAH65675MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH65675MD 250 mg
AAH6567503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH65675MD Supplier Thermo Scientific Chemicals Supplier No. H65675MD

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56564-52-4
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.219
MDL Number MFCD00069560
InChI Key SCIFESDRCALIIM-SECBINFHSA-N
Synonym r-2-methylamino-3-phenylpropanoic acid, n-methyl-d-phenylalanine, d-phenylalanine, n-methyl, 2r-2-methylamino-3-phenylpropanoic acid, h-d-mephe-oh hcl, ambotzhaa1199, l-phenylalanine, n-methyl
PubChem CID 6427037
IUPAC Name (2R)-2-(methylamino)-3-phenylpropanoic acid
SMILES CNC(CC1=CC=CC=C1)C(=O)O

Specifications

Beilstein 3199933
Quantity 250 mg
Formula Weight 179.22
Percent Purity 97%
Physical Form Solid
Chemical Name or Material N-Methyl-D-phenylalanine
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.