Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methyl-DL-alanine, 98%

Catalog No. AAH6528722
Click to view available options
Quantity:
25 g
100 g

CAS: 600-21-5 | C4H9NO2 | 103.121 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 600-21-5
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.121
MDL Number MFCD00063136
InChI Key GDFAOVXKHJXLEI-UHFFFAOYSA-N
Synonym n-methyl-dl-alanine, 2-methylamino propanoic acid, h-n-me-dl-ala-oh, n-alpha-methyl-dl-alanine, n-me-dl-ala-oh, alanine, n-methyl-, l, dl-n-methylalanine, me-dl-ala-oh, n-methyl cdl-alanine, n-, a-methyl-dl-alanine
PubChem CID 4377
IUPAC Name 2-(methylamino)propanoic acid
SMILES CC(C(=O)O)NC

Specifications

Color White
Quantity 100 g
Formula Weight 103.12
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material N-Methyl-DL-alanine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.