Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methyl-L-valine methyl ester hydrochloride, 95%, Thermo Scientific Chemicals

Catalog No. p-7056374
Click to view available options
Quantity:
250 mg
1 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3339-44-4
Molecular Formula C7H16ClNO2
Molecular Weight (g/mol) 181.66
MDL Number MFCD00238325
InChI Key DLLHGHUMLSLOID-RGMNGODLSA-N
Synonym n-me-val-ome hcl, n-methyl-l-valine methyl ester hydrochloride, n-me-val-ome . hcl, h-nmeval-ome hydrochloride, methyl n-methyl-l-valinate hydrochloride, n-methyl-l-valine methyl ester hydrochlorid, l-valine, n-methyl-, methyl ester, hydrochloride, methyl 2s-3-methyl-2-methylamino butanoate hydrochloride
PubChem CID 13819431
IUPAC Name methyl (2S)-3-methyl-2-(methylamino)butanoate hydrochloride
SMILES Cl.CN[C@@H](C(C)C)C(=O)OC

Specifications

Melting Point 103°C to 104°C
Beilstein 3562296
Quantity 250 mg
Formula Weight 181.66
Percent Purity 95%
Physical Form Solid
Chemical Name or Material N-Methyl-L-valine methyl ester hydrochloride
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.