Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methylaniline, 99%

Catalog No. p-3598429
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
Packaging:
Glass bottle

CAS: 100-61-8 | C7H9N | 107.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-61-8
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.16
MDL Number MFCD00008283
InChI Key AFBPFSWMIHJQDM-UHFFFAOYSA-N
Synonym methylaniline, monomethylaniline, n-methyl aniline, benzenamine, n-methyl, methylphenylamine, n-methylbenzenamine, n-monomethylaniline, n-methylaminobenzene, methylamino benzene, anilinomethane
PubChem CID 7515
ChEBI CHEBI:15733
IUPAC Name N-methylaniline
SMILES CNC1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -57.0°C
Color Brown to Yellow
Density 0.9870g/mL
Boiling Point 195.0°C
Flash Point 86°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5694 to 1.5714
Linear Formula C6H5NHCH3
Quantity 500 g
Beilstein 12, 135
Fieser 14,207
Specific Gravity 0.987
Merck Index 15, 6093
Solubility Information Solubility in water: 30g/l. Other solubilities: soluble in alcohol and ether,soluble in acetone,diethyl ether,ethanol and,chloroform
Formula Weight 107.15
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material N-Methylaniline
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
May cause damage to organs through prolonged or repeated exposure.
Toxic if inhaled.
Toxic in contact with skin.
Very toxic to aquatic life with long lasting effects.
Toxic if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 202-870-9

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.