Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
N-Methylcyclohexylamine 99.0+%, TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Click to view available options
Quantity:
25 mL
500 mL
Specifications
Specifications
Color | Colorless |
Boiling Point | 148°C |
Quantity | 25 mL |
UN Number | 2734 |
Formula Weight | 113.20 |
Percent Purity | ≥99.0% (T) |
Physical Form | Liquid |
Chemical Name or Material | N-Methylcyclohexylamine |
Safety and Handling
EINECSNumber | (3)-4003 |
RTECSNumber | GX1529000 |
TSCA | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction