Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methylformanilide, 99%

Catalog No. AC126810050
Click to view available options
Quantity:
5 g
100 g
500 g
1 kg
10 kg
Packaging:
Glass Bottle
Plastic drum

CAS: 93-61-8 | C8H9NO | 135.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-61-8
MDL Number MFCD00003283
InChI Key JIKUXBYRTXDNIY-UHFFFAOYSA-N
Synonym n-methylformanilide, formamide, n-methyl-n-phenyl, n-formyl-n-methylaniline, formanilide, n-methyl, methylphenylformamide, n-phenyl-n-methylformamide, n-methyl-n-formylaniline, n-methylformanilid, methyl phenyl formamide, n-methyl-n-phenyl-formamide
PubChem CID 66737
IUPAC Name N-methyl-N-phenylformamide
SMILES CN(C=O)C1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 8.0°C to 13.0°C
Color Colorless to Yellow
Density 1.0900g/mL
Boiling Point 243.0°C to 244.0°C
Flash Point 126°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Molecular Formula C8H9NO
Refractive Index 1.5600 to 1.5620
Linear Formula HCON(CH3)C6H5
Quantity 5 g
Specific Gravity 1.09
Solubility Information Solubility in water: immiscible.
Molecular Weight (g/mol) 135.166
Formula Weight 135.17
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material N-Methylformanilide, 99%
Show More Show Less
Hazard Statement GHS H Statement
Harmful if swallowed.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
EINECSNumber 202-262-3
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.