Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methylisobutylamine, 98+%

Catalog No. p-7058649
Click to view available options
Quantity:
1 g
5 g

CAS: 625-43-4 | C5H13N | 87.166 g/mol

N-Methylisobutylamine is used as an pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 625-43-4
Molecular Formula C5H13N
Molecular Weight (g/mol) 87.166
MDL Number MFCD00015043
InChI Key QKYWADPCTHTJHQ-UHFFFAOYSA-N
Synonym n-methylisobutylamine, 1-propanamine, n,2-dimethyl, n,2-dimethylpropylamine, n-isobutylmethylamine, methyl 2-methylpropyl amine, isobutylmethylamine, methylisobutylamine, propylamine, n,2-dimethyl, n-isobutyl-n-methylamine, methyl-isobutyl-amin
PubChem CID 12249
IUPAC Name N,2-dimethylpropan-1-amine
SMILES CC(C)CNC

Specifications

Density 0.73 g/mL
Boiling Point 78°C
Odor Amine-like
Quantity 1 g
UN Number UN2733
Solubility Information Miscible with water.
Formula Weight 87.16
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material N-Methylisobutylamine
Hazard Category H225-H314-H335
Hazard Statement GHS H Statement
H224-H314-H318
Extremely flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber 210-893-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.