Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methylmaleimide, 97%

Catalog No. p-3736943
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 930-88-1 | C5H5NO2 | 111.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 930-88-1
Molecular Formula C5H5NO2
Molecular Weight (g/mol) 111.1
MDL Number MFCD00005508
InChI Key SEEYREPSKCQBBF-UHFFFAOYSA-N
Synonym n-methylmaleimide, 1-methyl-1h-pyrrole-2,5-dione, maleimide, n-methyl, n-methylmaleinimide, 1h-pyrrole-2,5-dione, 1-methyl, n-methyl maleimide, unii-p0tfz8r21y, p0tfz8r21y, 1-methyl-pyrrole-2,5-dione, 1-methyl-2,5-dihydro-1h-pyrrole-2,5-dione
PubChem CID 70261
IUPAC Name 1-methylpyrrole-2,5-dione
SMILES CN1C(=O)C=CC1=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 93.0°C to 98.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Beilstein 21, 399
Formula Weight 111.1
Percent Purity 97%
Physical Form Crystals
Chemical Name or Material N-Methylmaleimide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal if swallowed.
May cause an allergic skin reaction.
Toxic if inhaled.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:
EINECSNumber 213-226-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.