Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sigma Aldrich N-Methylpyrrolidine
Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
5 mL
100 mL
500 mL
Description
- N-methylpyrrolidine can be used as a starting material to synthesize: n-methyl pyrrolidine-zinc borohydride (zbhnmp), a reducing agent for reduction of aldehydes, ketones, acid chlorides, and esters
- Zbhnmp is also used in the reductive amination of aldehydes and ketones to their corresponding amines.n-alkyl-n-methyl-pyrrolidinium bis(trifluoromethanesulfonyl)imide ionic liquids.n-methylpyrrolidine-2-one hydrotribromide applicable as a catalyst for aziridination of alkenes
Specifications
Specifications
| CAS | 120-94-5 |
| Density | 0.819 g/mL (at 25°C (literature)) |
| Boiling Point | 80°C to 81°C (lit.) |
| Molecular Formula | C5H11N |
| Linear Formula | C5H11N |
| MDL Number | MFCD00003173 |
| Quantity | 500 mL |
| Molecular Weight (g/mol) | 85.15 |
| Percent Purity | 97% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction