Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methylsuccinimide 98.0+%, TCI America™
SDP

Catalog No. M220725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M220725G 25 g
M22075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M220725G Supplier TCI America Supplier No. M220725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1121-07-9
Molecular Formula C5H7NO2
Molecular Weight (g/mol) 113.12
MDL Number MFCD00005517
InChI Key KYEACNNYFNZCST-UHFFFAOYSA-N
Synonym n-methylsuccinimide, 2,5-pyrrolidinedione, 1-methyl, succinimide, n-methyl, 1-methyl-2,5-pyrrolidinedione, n-methyl succinimide, acmc-2099cm, 1-methylazolidine-2,5-dione, ksc492s6n, 1-methyl-pyrrolidine-2,5-dione
PubChem CID 70717
IUPAC Name 1-methylpyrrolidine-2,5-dione
SMILES CN1C(=O)CCC1=O

Specifications

Melting Point 70°C
Color White
Boiling Point 235°C
Quantity 25 g
Formula Weight 113.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-Methylsuccinimide
RTECSNumber UY1028650
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.