Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methylthiourea 97%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
598-52-7
C2H6N2S
90.14
MFCD00004938
KQJQICVXLJTWQD-UHFFFAOYSA-N
n-methylthiourea,1-methylthiourea,1-methyl-2-thiourea,methylthiocarbamide,thiourea, methyl,monomethylthiourea,1-methylthiocarbamide,methyl thiourea,n-methylthiocarbamide,urea, 1-methyl-2-thio
2723704
methylthiourea
CNC(=S)N
598-52-7
C2H6N2S
90.14
MFCD00004938
KQJQICVXLJTWQD-UHFFFAOYSA-N
n-methylthiourea,1-methylthiourea,1-methyl-2-thiourea,methylthiocarbamide,thiourea, methyl,monomethylthiourea,1-methylthiocarbamide,methyl thiourea,n-methylthiocarbamide,urea, 1-methyl-2-thio
2723704
methylthiourea
CNC(=S)N
Melting Point 118°C to 123°C
Quantity 100g
Packaging Glass bottle
Infrared Spectrum Authentic
Color White to Yellow
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula CH3NHCSNH2
Beilstein 04, 70
Solubility Information Solubility in water: 165g/L (20°C). Other solubilities: 25g/L isopropanol (20°C),insoluble in non-polar solvents
Formula Weight 90.14
Physical Form Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material N-Methylthiourea, 97%
Show More Show Less

GHS H Statement:
Toxic if swallowed.
Suspected of damaging the unborn child.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 209-936-6

RTECSNumber : YT9000000