Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
n-Myc Antibody (NMYC-1) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 5 publications
$207.00 - $499.50
Specifications
Antigen | n-Myc |
---|---|
Clone | NMYC-1 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:500 - 1:1000, Chromatin Immunoprecipitation 1:10 - 1:500. Use reported in scientific literature (PMID 21068393), Flow Cytometry reported in scientific literature (PMID 27539729), Immunocytochemistry/Immunofluorescence 1:50 - 1:100, Immunoprecipitation 1:10 - 1:100, Gel Super Shift Assays 1:1 - 1:100. Use reported in scientific literature (PMID 21068393), Chromatin Immunoprecipitation (ChIP) 1:10-1:500, CyTOF-ready |
Applications | Western Blot, ChIP Assay, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB200109SS
![]() |
Novus Biologicals
NB200109SS |
0.025 mL |
Each for $207.00
|
|
|||||
NB200109
![]() |
Novus Biologicals
NB200109 |
0.1 mL |
Each for $499.50
|
|
|||||
Description
n-Myc Monoclonal specifically detects n-Myc in Human, Mouse samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Gel Super Shift Assays, CyTOF-ready.Specifications
n-Myc | |
1.0 mg/mL | |
Western Blot, ChIP Assay, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Cancer, Cell Cycle and Replication, Transcription Factors and Regulators, Tumor Suppressors | |
P04198 | |
4613 | |
A synthetic peptide (SPYVESEDAPPQKC) conjugated to KLH, corresponding to amino acids 327-339 of human n-Myc. [UniProt# P04198] | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
NMYC-1 | |
Western Blot 1:500 - 1:1000, Chromatin Immunoprecipitation 1:10 - 1:500. Use reported in scientific literature (PMID 21068393), Flow Cytometry reported in scientific literature (PMID 27539729), Immunocytochemistry/Immunofluorescence 1:50 - 1:100, Immunoprecipitation 1:10 - 1:100, Gel Super Shift Assays 1:1 - 1:100. Use reported in scientific literature (PMID 21068393), Chromatin Immunoprecipitation (ChIP) 1:10-1:500, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
BHLHE37, bHLHe37N-myc proto-oncogene protein, Class E basic helix-loop-helix protein 37, MODED, neuroblastoma-derived v-myc avian myelocytomatosis viral related oncogene, N-myc, NMYCneuroblastoma MYC oncogene, ODED, oncogene NMYC, pp65/67, v-myc avian myelocytomatosis viral related oncogene, neuroblastoma derived, v-myc myelocytomatosis viral related oncogene, neuroblastoma derived (avian) | |
MYCN | |
IgG2a κ | |
Protein A purified | |
This antibody does not react with c-Myc or L-Myc. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title