Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
N myc interactor Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100763AF750
Description
N myc interactor Polyclonal antibody specifically detects N myc interactor in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
N myc interactor | |
Polyclonal | |
Western Blot, Immunohistochemistry | |
Nmi, N-myc (and STAT) interactor, N-myc and STAT interactor, N-myc interactor, N-myc-interactor | |
The immunogen recognized by this antibody maps to a region between residues 1 and 50 of human N-myc and STAT interactor using the numbering given in entry NP_004679.1 (GeneID 9111). | |
0.1 mL | |
Cancer, Signal Transduction, Tumor Suppressors | |
9111 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction