Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
N myc interactor Antibody (PCRP-NMI-1C1), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308812FR
Description
N myc interactor Monoclonal antibody specifically detects N myc interactor in Human samples. It is validated for Flow Cytometry, Protein ArraySpecifications
N myc interactor | |
Monoclonal | |
DyLight 680 | |
Nmi, N-myc (and STAT) interactor, N-myc and STAT interactor, N-myc interactor, N-myc-interactor | |
Recombinant full-length human N myc interactor protein (Uniprot: Q13287) | |
100 μg | |
Cancer, Signal Transduction, Tumor Suppressors | |
9111 | |
Store at 4°C in the dark. | |
IgG2b |
Flow Cytometry, Protein Array | |
PCRP-NMI-1C1 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction