Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
n-Myc Antibody (NMYC-1) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP280882
Description
n-Myc Monoclonal specifically detects n-Myc in Human, Mouse samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Gel Super Shift Assays, CyTOF-ready.Specifications
| n-Myc | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot : 1:500 - 1:1000, Chromatin Immunoprecipitation 1:10 - 1:500. Use reported in scientific literature (PMID 21068393), Flow Cytometry reported in scientific literature (PMID 27539729), Immunocytochemistry/Immunofluorescence : 1:50 - 1:100, Immunoprecipitation : 1:10 - 1:100, Gel Super Shift Assays 1:1 - 1:100. Use reported in scientific literature (PMID 21068393), Chromatin Immunoprecipitation (ChIP) 1:10-1:500, CyTOF-ready | |
| BHLHE37, bHLHe37N-myc proto-oncogene protein, Class E basic helix-loop-helix protein 37, MODED, neuroblastoma-derived v-myc avian myelocytomatosis viral related oncogene, N-myc, NMYCneuroblastoma MYC oncogene, ODED, oncogene NMYC, pp65/67, v-myc avian myelocytomatosis viral related oncogene, neuroblastoma derived, v-myc myelocytomatosis viral related oncogene, neuroblastoma derived (avian) | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ChIP Assay, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| NMYC-1 | |
| Unconjugated | |
| PBS | |
| MYCN | |
| A synthetic peptide (SPYVESEDAPPQKC) conjugated to KLH, corresponding to amino acids 327-339 of human n-Myc. [UniProt# P04198] | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Phospho Specific, Transcription Factors and Regulators, Tumor Suppressors | |
| 4613 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction