Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Bis(4-bromophenyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline 98.0+%, TCI America™
SDP

Catalog No. B46601G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B46601G 1 g
B4660200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B46601G Supplier TCI America Supplier No. B46601G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 850153-24-1
Molecular Formula C24H24BBr2NO2
Molecular Weight (g/mol) 529.079
InChI Key XVOQIMZHMJQJEC-UHFFFAOYSA-N
Synonym 2-[4-[Bis(4-bromophenyl)amino]phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-[Bis(4-bromophenyl)amino]phenylboronic Acid Pinacol Ester
PubChem CID 89535548
IUPAC Name N,N-bis(4-bromophenyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)N(C3=CC=C(C=C3)Br)C4=CC=C(C=C4)Br

Specifications

Melting Point 149°C
Color White
Quantity 1 g
Formula Weight 529.08
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N,N-Bis(4-bromophenyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.