Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Bis[4-(diphenylamino)phenyl]-N,N'-diphenylbenzidine 98.0+%, TCI America™
SDP

Catalog No. B51171G
Click to view available options
Quantity:
200 mg
1 g

Chemical Identifiers

CAS 209980-53-0
Molecular Formula C60H46N4
Molecular Weight (g/mol) 823.06
MDL Number MFCD28015747
InChI Key XOYZGLGJSAZOAG-UHFFFAOYSA-N
PubChem CID 21918000
IUPAC Name N4,N4'-bis[4-(diphenylamino)phenyl]-N4,N4'-diphenyl-[1,1'-biphenyl]-4,4'-diamine
SMILES C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 250°C
Quantity 1 g
Formula Weight 823.06
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Bis[4-(diphenylamino)phenyl]-N,N′-diphenylbenzidine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.