Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Bis(4-methoxy-2-methylphenyl)-N,N'-diphenylbenzidine 98.0+%, TCI America™
SDP

Catalog No. B447025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B447025G 25 g
B44705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B447025G Supplier TCI America Supplier No. B447025G

Chemical Identifiers

CAS 169685-34-1
Molecular Formula C40H36N2O2
Molecular Weight (g/mol) 576.74
MDL Number MFCD28384132
InChI Key HACXFZALVVMGPO-UHFFFAOYSA-N
PubChem CID 23517938
IUPAC Name 4-methoxy-N-[4-[4-(N-(4-methoxy-2-methylphenyl)anilino)phenyl]phenyl]-2-methyl-N-phenylaniline
SMILES CC1=C(C=CC(=C1)OC)N(C2=CC=CC=C2)C3=CC=C(C=C3)C4=CC=C(C=C4)N(C5=CC=CC=C5)C6=C(C=C(C=C6)OC)C

Specifications

Melting Point 160°C
Color Yellow
Quantity 25 g
Formula Weight 576.74
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Bis(4-methoxy-2-methylphenyl)-N,N′-diphenylbenzidine
EINECSNumber (4)-1917
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.