Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Di(4-pyridyl)-1,4,5,8-naphthalenetetracarboxdiimide 98.0+%, TCI America™
SDP

Catalog No. p-7128128
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 34151-49-0
Molecular Formula C24H12N4O4
Molecular Weight (g/mol) 420.384
InChI Key IBRPEOCBRYYINT-UHFFFAOYSA-N
PubChem CID 1381898
SMILES C1=CC2=C3C(=CC=C4C3=C1C(=O)N(C4=O)C5=CC=NC=C5)C(=O)N(C2=O)C6=CC=NC=C6

Specifications

Melting Point 353°C
Quantity 1 g
Formula Weight 420.38
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Di(4-pyridyl)-1,4,5,8-naphthalenetetracarboxdiimide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.