Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Dibenzylethylenediamine diacetate, 99%

Catalog No. aal1797414
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1797414 25 g
AAL1797422 100 g
AAL1797436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1797414 Supplier Thermo Scientific Chemicals Supplier No. L1797414
Only null left
Add to Cart

CAS: 122-75-8 | C20H28N2O4 | 360.454 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-75-8
Molecular Formula C20H28N2O4
Molecular Weight (g/mol) 360.454
MDL Number MFCD00040588
InChI Key MTRNNCLQPVCDLF-UHFFFAOYSA-N
Synonym n,n'-dibenzylethylenediamine diacetate, n1,n2-dibenzylethane-1,2-diamine diacetate, n,n'-dibenzyl ethylenediamine diacetate, benzathine diacetate, dbed diacetate, unii-x18b8x2c22, n,n'-dibenzylethylenediammonium di acetate, 1,2-ethanediamine, n,n'-bis phenylmethyl-, diacetate, bis acetic acid ; benzathine, 1,2-di benzylamino ethane diacetate
PubChem CID 31228
IUPAC Name acetic acid;N,N'-dibenzylethane-1,2-diamine
SMILES CC(=O)O.CC(=O)O.C1=CC=C(C=C1)CNCCNCC2=CC=CC=C2

Specifications

Melting Point 117°C to 118°C
Quantity 25 g
Beilstein 3781362
Merck Index 14,1064
Formula Weight 360.46
Percent Purity 99%
Chemical Name or Material N,N'-Dibenzylethylenediamine diacetate
Hazard Category H302
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 204-572-4
RTECSNumber KV3400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.