Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dibenzylhydrazine 96.0+%, TCI America™
SDP

Catalog No. D2132500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2132500G 500 g
D213225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D2132500G Supplier TCI America Supplier No. D2132500G

Chemical Identifiers

CAS 5802-60-8
Molecular Formula C14H16N2
Molecular Weight (g/mol) 212.296
MDL Number MFCD00144976
InChI Key CZVXEAWVGZTLON-UHFFFAOYSA-N
PubChem CID 197050
IUPAC Name 1,1-dibenzylhydrazine
SMILES C1=CC=C(C=C1)CN(CC2=CC=CC=C2)N

Specifications

Melting Point 56°C
Color White-Yellow
Boiling Point 171°C
Quantity 500 g
Formula Weight 212.30
Percent Purity ≥96.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material N,N-Dibenzylhydrazine
RTECSNumber MV2086000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.