Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Dicyclohexylcarbodiimide, 99%

Catalog No. p-7081141
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 538-75-0 | C13H22N2 | 206.333 g/mol

N,N'-Dicyclohexylcarbodiimide is a carboxyl activating reagent used in peptide synthesis. It acts as a dehydrating agent for the preparation of amides, ketones and nitriles. It is involved in the preparation of esters by reacting tertiary alcohols with carboxylic acid. It is utilized for the stereo chemical inversion of secondary alcohols by forming formyl ester followed by saponification. It is associated with dimethyl sulfoxide and used in Pfitzner-Moffatt oxidation to convert alcohols into aldehydes and ketones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N′-Dicyclohexylcarbodiimide is a carboxyl activating reagent used in peptide synthesis. It acts as a dehydrating agent for the preparation of amides, ketones and nitriles. It is involved in the preparation of esters by reacting tertiary alcohols with carboxylic acid. It is utilized for the stereo chemical inversion of secondary alcohols by forming formyl ester followed by saponification. It is associated with dimethyl sulfoxide and used in Pfitzner-Moffatt oxidation to convert alcohols into aldehydes and ketones.

Solubility
Soluble in methanol, chloroform, dichloromethane, acetonitrile, tetrahydrofuran and dimethylformamide. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 538-75-0
Molecular Formula C13H22N2
Molecular Weight (g/mol) 206.333
MDL Number MFCD00011659
InChI Key QOSSAOTZNIDXMA-UHFFFAOYSA-N
Synonym dicyclohexylcarbodiimide, n,n'-dicyclohexylcarbodiimide, dccd, 1,3-dicyclohexylcarbodiimide, dcci, carbodicyclohexylimide, bis cyclohexyl carbodiimide, dcc, n,n-dicyclohexylcarbodiimide, carbodiimide, dicyclohexyl
PubChem CID 10868
ChEBI CHEBI:53090
IUPAC Name N,N'-dicyclohexylmethanediimine
SMILES C1CCC(CC1)N=C=NC2CCCCC2

Specifications

Melting Point 33°C to 36°C
Boiling Point 154°C to 156°C (11 mmHg)
Flash Point 138°C (280°F)
Quantity 25 g
UN Number UN2811
Beilstein 610662
Sensitivity Moisture sensitive
Merck Index 14,3096
Solubility Information Soluble in methanol,chloroform,dichloromethane,acetonitrile,tetrahydrofuran and dimethylformamide. Insoluble in water.
Formula Weight 206.33
Percent Purity 99%
Chemical Name or Material N,N'-Dicyclohexylcarbodiimide
Show More Show Less
Hazard Category H302-H311-H317-H318
Hazard Statement GHS H Statement
H311-H318-H302-H317
Toxic in contact with skin.
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P310-P312-P330-P333+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 208-704-1
RTECSNumber FF2160000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.