Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Dicyclohexylcarbodiimide, 99%

Catalog No. AC113900250
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
5 kg
25 kg
Packaging:
Glass bottle
Plastic drum
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC113900250 25 kg Plastic drum
AC113901000 100 g Glass bottle
AC113902500 250 g Glass bottle
AC113900010 1 kg Glass bottle
AC113900050 5 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. AC113900250 Supplier Thermo Scientific Chemicals Supplier No. 113900250
Add to Cart

N,N-Dicyclohexylcarbodiimide, 99%, C13H22N2, CAS Number-538-75-0, carbodiimide, dicyclohexyl, bis cyclohexyl carbodiimide, dcc, 1,3-dicyclohexylcarbodiimide, carbodicyclohexylimide, n,n'-dicyclohexylcarbodiimide, n,n-dicyclohexylcarbodiimide, dcci, dicyclohexylcarbodiimide, dccd, 1kg | CAS: 538-75-0 | C13H22N2 | 206.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 538-75-0
Molecular Formula C13H22N2
Molecular Weight (g/mol) 206.33
MDL Number MFCD00011659
InChI Key QOSSAOTZNIDXMA-UHFFFAOYSA-N
Synonym dicyclohexylcarbodiimide, n,n'-dicyclohexylcarbodiimide, dccd, 1,3-dicyclohexylcarbodiimide, dcci, carbodicyclohexylimide, bis cyclohexyl carbodiimide, dcc, n,n-dicyclohexylcarbodiimide, carbodiimide, dicyclohexyl
PubChem CID 10868
ChEBI CHEBI:53090
IUPAC Name N,N'-dicyclohexylmethanediimine
SMILES C1CCC(CC1)N=C=NC2CCCCC2

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 33.0°C to 35.0°C
Color White to Yellow
Boiling Point 122.0°C to 124.0°C (6.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic drum
Linear Formula C6H11N=C=NC6H11
Quantity 25 kg
UN Number 2928
Fieser 01,231; 02,126; 03,91; 04,141; 05,206; 06,174; 07,100; 08,162; 09,156; 10,142; 11,173; 13,333; 14,131; 15,131; 16,128
Merck Index 15,3107
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents,100g/l dichloromethane
Viscosity 5.81 mPa.s (55°C)
Formula Weight 206.33
Percent Purity 99%
Physical Form Crystalline Mass or Waxy Solid
Chemical Name or Material N,N′-Dicyclohexylcarbodiimide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic in contact with skin.
Harmful if swallowed.
Causes serious eye damage.
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water
EINECSNumber 208-704-1

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.