Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl aminomalonate hydrochloride, 98%

Catalog No. AC114041000
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle

CAS: 13433-00-6 | C7H13NO4·ClH | 211.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13433-00-6
Molecular Formula C7H13NO4·ClH
Molecular Weight (g/mol) 211.64
MDL Number MFCD00012510
InChI Key GLFVNTDRBTZJIY-UHFFFAOYSA-N
Synonym diethyl aminomalonate hydrochloride, diethyl 2-aminomalonate hydrochloride, aminomalonic acid diethyl ester hydrochloride, 1,3-diethyl 2-aminopropanedioate hydrochloride, propanedioic acid, amino-, diethyl ester, hydrochloride, diethyl 2-aminopropanedioate hydrochloride, diethyl 2-aminomalonate hcl, diethyl aminomalonate hcl, diethyl 2-aminopropane-1,3-dioate, chloride, pubchem23977
PubChem CID 3084143
IUPAC Name diethyl 2-aminopropanedioate;hydrochloride
SMILES CCOC(=O)C(C(=O)OCC)N.Cl

Specifications

Melting Point 166.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula C2H5O2CCH(NH2)CO2C2H5·HCl
Quantity 100 g
Beilstein 04, 470
Fieser 15,132
Solubility Information Solubility in water: soluble
Formula Weight 211.64
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Diethyl aminomalonate hydrochloride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 236-556-8

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.