Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Diethyl-1,4-phenylenediamine Sulfate 98.0+%, TCI America™
SDP

Catalog No. D0520100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0520100G 100 g
D052025G 25 g
D0520500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D0520100G Supplier TCI America Supplier No. D0520100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6283-63-2
Molecular Formula C10H18N2O4S
Molecular Weight (g/mol) 262.324
MDL Number MFCD00012993
InChI Key AYLDJQABCMPYEN-UHFFFAOYSA-N
Synonym n,n-diethyl-p-phenylenediamine sulfate, n1,n1-diethylbenzene-1,4-diamine sulfate, 4-amino-n,n-diethylaniline sulfate, diethyl-p-phenylenediamine sulfate, n,n-diethyl-1,4-phenylenediamine sulfate, unii-usp19t3gda, 1,4-benzenediamine, n,n-diethyl-, sulfate 1:1, 1,4-benzenediamine, n,n-diethyl-, sulfate, n,n-diethyl-1,4-benzenediamine sulfate, p-phenylenediamine, n,n-diethyl-, sulfate 1:1
PubChem CID 80166
IUPAC Name 4-N,4-N-diethylbenzene-1,4-diamine;sulfuric acid
SMILES CCN(CC)C1=CC=C(C=C1)N.OS(=O)(=O)O

Specifications

Melting Point 185°C
Quantity 100 g
Formula Weight 262.32
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N,N-Diethyl-1,4-phenylenediamine Sulfate
EINECSNumber (3)-0243
RTECSNumber SS9625000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.