Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Diethyl-1,6-diaminohexane 97.0+%, TCI America™
SDP

Catalog No. D130525ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D130525ML 25 mL
D13055ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D130525ML Supplier TCI America Supplier No. D130525ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13093-05-5
Molecular Formula C10H24N2
Molecular Weight (g/mol) 172.32
MDL Number MFCD00026707
InChI Key LDQWVRMGQLAWMN-UHFFFAOYSA-N
PubChem CID 83132
IUPAC Name ethyl[6-(ethylamino)hexyl]amine
SMILES CCNCCCCCCNCC

Specifications

Color Yellow
Boiling Point 228°C
Quantity 25 mL
UN Number 2735
Synonym 1,6-Bis(ethylamino)hexane, N,N′-Diethylhexamethylenediamine, N,N′-Diethyl-1,6-hexanediamine
Formula Weight 172.32
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N,N′-Diethyl-1,6-diaminohexane
EINECSNumber (2)-2374
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.