Learn More
N,N-Diethyl-3-methylbenzamide, 97%
Description
It is the most common ingredient in insect repellants and is believed to work as such in that mosquitoes intensely dislike its smell. Deet is also an effective solvent and may dissolve plastics, rayon, spandex, other synthetic fabrics and painted or varnished surfaces.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is the most common ingredient in insect repellants and is believed to work as such in that mosquitoes intensely dislike its smell. Deet is also an effective solvent and may dissolve plastics, rayon, spandex, other synthetic fabrics and painted or varnished surfaces.
Solubility
Soluble in water.
Notes
Keep away from strong oxidizing agents, strong acids, strong bases, strong reducing agents. Keep ate room temperature.
Specifications
Specifications
| Density | 0.997 |
| Boiling Point | 110°C to 112°C (1 mmHg) |
| Flash Point | 145°C (293°F) |
| Refractive Index | 1.523 |
| Quantity | 2500 g |
| Beilstein | 2046711 |
| Merck Index | 14,2856 |
| Solubility Information | Soluble in water. |
| Formula Weight | 191.27 |
| Percent Purity | 97% |
| Show More |
Safety and Handling
| Hazard Category | H302-H315-H319-H336-H373 |
| Hazard Statement | GHS H Statement H302-H315-H319 Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c |
| EINECSNumber | 205-149-7 |
| RTECSNumber | XS3675000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.