Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Diethyl-3-methylbenzamide, 97%

Catalog No. aaa1748622
Click to view available options
Quantity:
100 g
500 g
2500 g

CAS: 134-62-3 | C12H17NO | 191.274 g/mol

It is the most common ingredient in insect repellants and is believed to work as such in that mosquitoes intensely dislike its smell. Deet is also an effective solvent and may dissolve plastics, rayon, spandex, other synthetic fabrics and painted or varnished surfaces.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is the most common ingredient in insect repellants and is believed to work as such in that mosquitoes intensely dislike its smell. Deet is also an effective solvent and may dissolve plastics, rayon, spandex, other synthetic fabrics and painted or varnished surfaces.

Solubility
Soluble in water.

Notes
Keep away from strong oxidizing agents, strong acids, strong bases, strong reducing agents. Keep ate room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 134-62-3
Molecular Formula C12H17NO
Molecular Weight (g/mol) 191.274
MDL Number MFCD00009046
InChI Key MMOXZBCLCQITDF-UHFFFAOYSA-N
Synonym deet, n,n-diethyl-m-toluamide, diethyltoluamide, dieltamid, metadelphene, flypel, delphene, deta, detamide, autan
PubChem CID 4284
ChEBI CHEBI:7071
IUPAC Name N,N-diethyl-3-methylbenzamide
SMILES CCN(CC)C(=O)C1=CC(=CC=C1)C

Specifications

Density 0.997
Boiling Point 110°C to 112°C (1 mmHg)
Flash Point 145°C (293°F)
Refractive Index 1.523
Quantity 100 g
Beilstein 2046711
Merck Index 14,2856
Solubility Information Soluble in water.
Formula Weight 191.27
Percent Purity 97%
Chemical Name or Material N,N-Diethyl-3-methylbenzamide
Show More Show Less
Hazard Category H302-H315-H319-H336-H373
Hazard Statement GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
EINECSNumber 205-149-7
RTECSNumber XS3675000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.