Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Diethylacetoacetamide, 97%

Catalog No. AAB2401918
Click to view available options
Quantity:
50 g
250 g

Catalog No. AAB2401918

Supplier: Thermo Scientific Chemicals B2401918

Only null left
Add to Cart

CAS: 2235-46-3 | C8H15NO2 | 157.213 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2235-46-3
Molecular Formula C8H15NO2
Molecular Weight (g/mol) 157.213
MDL Number MFCD00026728
InChI Key NTMXFHGYWJIAAE-UHFFFAOYSA-N
Synonym n,n-diethylacetoacetamide, n,n-diethyl-3-oxobutyramide, diethylacetoacetamide, butanamide, n,n-diethyl-3-oxo, acetoacetamide, n,n-diethyl, 1-diethylcarbamoyl-2-propanone, n,n-diethylacetylacetamide, acetoacetdiethylamide, diethylamid kyseliny acetoctove, diethylamid kyseliny acetoctove czech
PubChem CID 16699
IUPAC Name N,N-diethyl-3-oxobutanamide
SMILES CCN(CC)C(=O)CC(=O)C

Specifications

Melting Point <-73°C
Density 0.99
Boiling Point 213°C to 218°C
Flash Point 94°C (201°F)
Refractive Index 1.472
Quantity 50 g
Beilstein 636486
Formula Weight 157.2
Percent Purity 97%
Chemical Name or Material N,N-Diethylacetoacetamide
EINECSNumber 218-792-3
RTECSNumber AK4025000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.