Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Diethylethylenediamine, 98+%, ACROS Organics™

Click to view available options
Quantity:
100mL
500mL
Packaging:
Glass bottle
100-36-7
C6H16N2
116.21
MFCD00008176
UDGSVBYJWHOHNN-UHFFFAOYSA-N
n,n-diethylethylenediamine,2-diethylamino ethylamine,n,n-diethyl-1,2-ethanediamine,diethylamino ethylamino,n1,n1-diethylethane-1,2-diamine,2-aminoethyl diethylamine,2-aminoethyldiethylamine,n,n-diethylethanediamine,ethylenediamine, n,n-diethyl,n,n-diethylethane-1,2-diamine
60993
N',N'-diethylethane-1,2-diamine
CCN(CC)CCN
100-36-7
C6H16N2
116.21
MFCD00008176
UDGSVBYJWHOHNN-UHFFFAOYSA-N
n,n-diethylethylenediamine,2-diethylamino ethylamine,n,n-diethyl-1,2-ethanediamine,diethylamino ethylamino,n1,n1-diethylethane-1,2-diamine,2-aminoethyl diethylamine,2-aminoethyldiethylamine,n,n-diethylethanediamine,ethylenediamine, n,n-diethyl,n,n-diethylethane-1,2-diamine
60993
N',N'-diethylethane-1,2-diamine
CCN(CC)CCN
Refractive Index 1.4350 to 1.437
Boiling Point 145°C to 147°C
Flash Point 30°C
Specific Gravity 0.82
Packaging Glass bottle
Water 0.5% max. (K.F.)
Color Colorless to Yellow
Quantity 500mL
Infrared Spectrum Authentic
Density 0.8200g/mL
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Linear Formula (C2H5)2NCH2CH2NH2
Beilstein 04, 251
Solubility Information Solubility in water: miscible. Other solubilities: soluble in ether,ethanol,toluene,ccl4
Formula Weight 116.21
Physical Form Liquid
Percent Purity ≥98.0%
Chemical Name or Material N, N-Diethylethylenediamine, 98+%
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 202-844-7

RTECSNumber : KV3500000

TSCA : TSCA