Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Diethylhydrazine dihydrochloride, tech. 90%

Catalog No. p-7026354
Click to view available options
Quantity:
5 g
25 g

CAS: 7699-31-2 | C4H14Cl2N2 | 161.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7699-31-2
Molecular Formula C4H14Cl2N2
Molecular Weight (g/mol) 161.07
MDL Number MFCD00012523
InChI Key NMTDWUVFGOBBAX-UHFFFAOYSA-N
Synonym 1,2-diethylhydrazine dihydrochloride, n,n'-diethylhydrazine dihydrochloride, unii-x3o3iwe4tg, ccris 8919, hydrazine, 1,2-diethyl-, dihydrochloride, x3o3iwe4tg, 1,2-diethylhydrazine 2hcl, diethylhydrazine dihydrochloride, 1,2-diethyl hydrazine dihydrochloride, 1,2-diethylhydrazine dihydrochlo-ride
PubChem CID 24372
IUPAC Name 1,2-diethylhydrazine;dihydrochloride
SMILES [H+].[H+].[Cl-].[Cl-].CCNNCC

Specifications

Melting Point ∼169°C (decomposition)
Odor Odorless
Quantity 5 g
Beilstein 3668380
Formula Weight 159.06
Percent Purity 90%
Grade Technical
Chemical Name or Material N,N'-Diethylhydrazine dihydrochloride
Hazard Category H302+H312-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
RTECSNumber MV2295000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.